Invitrogen
Manufacturer Code:PA5101116
Catalog # PIPA5101116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HOXA1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BSAS early retinoic acid 1 ERA1 homeo box A1 homeobox 1F homeobox A1 homeobox protein Hox-1.6 homeobox protein Hox-1F Homeobox protein Hox-A1 homeoboxless protein ERA-1-399 homeotic protein ERA-1-993 Hox 1.6-like protein HOX A1 homeodomain protein HOX1 Hox-1.6 HOX1F HOXA 1 Hoxa1 HOXA-1 lab-like protein MGC45232 NEWGENE_2812; early retinoic acid 1; homeo box A1; homeobox 1F; homeobox A1; homeobox protein Hox-1.6; Homeobox protein Hox-1F; Homeobox protein Hox-A1; homeoboxless protein ERA-1-399; homeotic protein ERA-1-993; Hox 1.6-like protein; HOX A1 homeodomain protein; lab-like protein
Gene Aliases: BSAS; ERA1; Hox-1.6; HOX1; HOX1F; HOXA1
UniProt ID: (Human) P49639
Entrez Gene ID: (Human) 3198, (Mouse) 15394, (Rat) 103690132
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.