R&D Systems
Manufacturer Code:AF5014
Catalog # AF5014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human HOXA1 Met1-Asp118 Accession # P49639 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BSAS homeo box A1 homeobox A1 Homeobox protein Hox-1F homeobox protein Hox-A1 Hox 1.6-like protein HOX A1 homeodomain protein HOX1 HOX1F HOX1Fhomeobox 1F lab-like protein MGC45232; homeo box A1; homeobox 1F; Homeobox protein Hox-1F; Homeobox protein Hox-A1; Hox 1.6-like protein; HOX A1 homeodomain protein; lab-like protein
Gene Aliases: BSAS; HOX1; HOX1F; HOXA1
UniProt ID: (Human) P49639
Entrez Gene ID: (Human) 3198
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.