Invitrogen
Manufacturer Code:PA543944
Catalog # PIPA543944
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human HORMAD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HORMA domain-containing protein 2
Gene Aliases: 4930529M09Rik; CT46.2; HORMAD2
UniProt ID: (Human) Q8N7B1, (Bovine) A6QQY4, (Mouse) Q5SQP1
Entrez Gene ID: (Horse) 100058792, (Human) 150280, (Rabbit) 100342243, (Bovine) 777596, (Dog) 611903, (Rat) 498400, (Mouse) 75828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.