Abnova
Manufacturer Code:PAB17406
Catalog # 89-119-063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human HORMAD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of HORMAD1.
For Research Use Only
Protein Aliases: Cancer/testis antigen 46; CT46; DKFZp434A1315; HORMA domain-containing protein 1; Newborn ovary HORMA protein; NOHMA; RP11-363I22.1; testis tissue sperm-binding protein Li 86P
Gene Aliases: CT46; HORMAD1; NOHMA
UniProt ID: (Human) Q86X24
Entrez Gene ID: (Human) 84072
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.