Proteintech
Manufacturer Code:139171AP150UL
Catalog # 50-172-9089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HORMAD1 Fusion Protein Ag4914 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 46; Cancer/testis antigen 46 CT46 HORMA domain containing 1 HORMAD1 Newborn ovary HORMA protein NOHMA RP11 363I22.1; CT46; HORMA domain containing protein; HORMA domain-containing protein 1; Newborn ovary HORMA protein; testis tissue sperm-binding protein Li 86P
Gene Aliases: 4921522K05Rik; CT46; HORMAD1; NOHMA; RGD1564960
UniProt ID: (Human) Q86X24, (Mouse) Q9D5T7, (Rat) D3ZWE7
Entrez Gene ID: (Human) 84072, (Mouse) 67981, (Rat) 365868
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.