Abnova
Manufacturer Code:H00084072K
Catalog # 89-208-736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human HORMAD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human HORMAD1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Cancer/testis antigen 46; CT46; DKFZp434A1315 NOHMA RP11-363I22.1; HORMA domain-containing protein 1; Newborn ovary HORMA protein; testis tissue sperm-binding protein Li 86P
Gene Aliases: CT46; HORMAD1; NOHMA
UniProt ID: (Human) Q86X24
Entrez Gene ID: (Human) 84072
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.