Proteintech
Manufacturer Code:114191AP150UL
Catalog # 50-172-7143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HOPX Fusion Protein Ag1979 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAMEO HOD Homeodomain only protein HOP HOP homeobox HOPX LAGY NECC1 OB1 Odd homeobox protein 1 SMAP31 TOTO; GIIg15b; global ischemia induced protein GIIG15B; Global ischemia-induced gene 15B protein; global ischemia-induced protein 15B; homeobox only domain; Homeobox-only protein; homeodomain only protein; Homeodomain-only protein; Lung cancer-associated Y protein; mOB1; not expressed in choriocarcinoma clone 1; Not expressed in choriocarcinoma protein 1; odd homeobox 1; Odd homeobox protein 1
Gene Aliases: 1110018K11Rik; 1200015P04Rik; 2300002F06Rik; AI848177; AW490897; CAMEO; GIIg15b; Hdop; HOD; HOP; HOPX; LAGY; NECC1; OB1; Obl; SMAP31; TOTO
UniProt ID: (Human) Q9BPY8, (Mouse) Q8R1H0, (Rat) Q78ZR5
Entrez Gene ID: (Human) 84525, (Mouse) 74318, (Rat) 171160
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.