Proteintech
Manufacturer Code:154571AP150UL
Catalog # 50-173-0243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HOOK3 Fusion Protein Ag7724 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: h hook3 hHK3 HK3 hook homolog 3 (Drosophila) HOOK3 Protein Hook homolog 3; h-hook3; hHK3; hook homolog 3; mHK3; Protein Hook homolog 3
Gene Aliases: 5830454D03Rik; AI317159; E330005F07Rik; HK3; HOOK3
UniProt ID: (Human) Q86VS8, (Mouse) Q8BUK6
Entrez Gene ID: (Human) 84376, (Rat) 306548, (Mouse) 320191
Molecular Function:
enzyme modulator
kinase modulator
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.