Novus Biologicals
Manufacturer Code:NBP270895G
Catalog # NBP270895G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C2 |
Immunogen | Full length recombinant protein of human HO-2/HMOX2 (NP_002125) produced in HEK293T cells. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: heme oxygenase (decycling) 2; heme oxygenase (decycling) 2 heme oxygenase 2 HO2 HO-2EC 1.14.99.3; Heme oxygenase 2; HO-2
Gene Aliases: HMOX2; HO-2; HO2
UniProt ID: (Human) P30519
Entrez Gene ID: (Human) 3163
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.