Proteintech
Manufacturer Code:272821AP150UL
Catalog # 50-173-5630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HO-1/HMOX1 Fusion Protein Ag26172 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bK286B10 heme oxygenase (decycling) 1 Heme oxygenase 1 HMOX1 HO HO 1 HO1 HO-1 HSP32; heat shock protein, 32-kD; heme oxygenase (decycling) 1; Heme oxygenase 1; hemoxygenase; HO-1; P32 protein
Gene Aliases: bK286B10; D8Wsu38e; Hemox; Hmox; HMOX1; HMOX1D; HO; HO-1; HO1; HSP32
UniProt ID: (Human) P09601, (Mouse) P14901
Entrez Gene ID: (Human) 3162, (Mouse) 15368
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.