Invitrogen
Manufacturer Code:PA579377
Catalog # PIPA579377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human HMOX1 recombit protein (Position: M1-M288). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: heat shock protein, 32-kD; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; Heme oxygenase 1; Heme oxygenase 1 HO-1; HO-1; HSP32
Gene Aliases: bK286B10; Heox; HEOXG; Hmox; HMOX1; HMOX1D; HO; HO-1; HO1; HSP32
UniProt ID: (Human) P09601, (Rat) P06762
Entrez Gene ID: (Human) 3162, (Rat) 24451
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.