Invitrogen
Manufacturer Code:PA577833
Catalog # PIPA577833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Human heme-oxygenase (HO-1) synthetic multiple antigenic peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: heat shock protein, 32-kD; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; Heme oxygenase 1; hemoxygenase; HO-1; HSP32; HSP32 HMOX1 Heme oxygenase 1 HO HO1 32 kD bK286B10 D8Wsu38e heat shock protein 32 kD heat shock protein 32kD Heat shock protein Heme oxygenase (decycling) 1 Heme oxygenase 1 Hemox HMOX 1 Hmox Hmox1 HMOX1&lowbarHUMAN HO 1 HO HO-1 HO1 Hsp32; P32 protein
Gene Aliases: bK286B10; D8Wsu38e; Hemox; Heox; HEOXG; Hmox; HMOX1; HMOX1D; HO; HO-1; HO1; HSP32
UniProt ID: (Human) P09601, (Rat) P06762, (Mouse) P14901
Entrez Gene ID: (Human) 3162, (Dog) 442987, (Rat) 24451, (Mouse) 15368
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.