Proteintech
Manufacturer Code:183541AP150UL
Catalog # 50-173-2132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HNRNPL Fusion Protein Ag13211 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Heterogeneous nuclear ribonucleoprotein L; heterogenous nuclear ribonucleoprotein L; hnRNP L; hnRNP L HNRNPL HNRPL P/OKcl.14
Gene Aliases: C79783; D830027H13Rik; Fblim1; hnRNP-L; HNRNPL; HNRPL; P/OKcl.14
UniProt ID: (Human) P14866, (Mouse) Q8R081, (Rat) F1LQ48
Entrez Gene ID: (Human) 3191, (Mouse) 15388, (Rat) 80846
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.