Proteintech
Manufacturer Code:108481AP150UL
Catalog # 50-172-6652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HNRNPA0 Fusion Protein Ag1312 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Heterogeneous nuclear ribonucleoprotein A0; hnRNA binding protein; hnRNP A0; hnRNP A0 HNRNPA0 HNRPA0
Gene Aliases: 1110055B05Rik; 3010025E17Rik; hnRNP A0; HNRNPA0; HNRPA0
UniProt ID: (Human) Q13151, (Mouse) Q9CX86
Entrez Gene ID: (Human) 10949, (Mouse) 77134
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.