Invitrogen
Manufacturer Code:PA557289
Catalog # PIPA557289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HNMT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Histamine N-methyltransferase; Histamine N-methyltransferase HMT HNMT; HMT
Gene Aliases: 1500031F01Rik; AI788969; HMT; HNMT; HNMT-S1; HNMT-S2; MRT51
UniProt ID: (Human) P50135, (Mouse) Q91VF2, (Rat) Q01984
Entrez Gene ID: (Human) 3176, (Mouse) 140483, (Rat) 81676
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.