Proteintech
Manufacturer Code:118741AP150UL
Catalog # 50-172-7526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HNMT Fusion Protein Ag2462 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: histamine N methyltransferase HMT HNMT HNMT S1 HNMT S2; Histamine N-methyltransferase; HMT
Gene Aliases: 1500031F01Rik; AI788969; HMT; HNMT; HNMT-S1; HNMT-S2; MRT51
UniProt ID: (Human) P50135, (Mouse) Q91VF2, (Rat) Q01984
Entrez Gene ID: (Human) 3176, (Mouse) 140483, (Rat) 81676
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.