Abnova
Manufacturer Code:PAB8769
Catalog # 89-133-922
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 454 of rat Hnf4a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Hnf4a.
For Research Use Only
Protein Aliases: alpha transcription factor 4; hepatic nuclear factor 4, alpha; Hepatocyte nuclear factor 4-alpha; HNF-4-alpha; Hnf4; Nuclear receptor subfamily 2 group A member 1; TCF-14; Tcf4; Transcription factor 14; Transcription factor HNF-4
Gene Aliases: Hnf-4; Hnf4; Hnf4a; Hnf4alpha; Nr2a1; Tcf14
UniProt ID: (Rat) P22449
Entrez Gene ID: (Rat) 25735
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.