Abnova
Manufacturer Code:MAB20171
Catalog # 89-336-571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | HOH-8 |
Immunogen | A synthetic peptide corresponding to human HNF1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: FJHN HNF1beta HNF2 HPC11 LF-B3 LFB3 MODY5 TCF2 VHNF1; Hepatocyte nuclear factor 1-beta; HNF-1-beta; HNF1 beta A; Homeoprotein LFB3; TCF-2; Transcription factor 2; transcription factor 2, hepatic; Variant hepatic nuclear factor 1; vHNF1
Gene Aliases: FJHN; HNF-1-beta; HNF-1B; HNF1B; HNF1beta; HNF2; HPC11; LF-B3; LFB3; MODY5; TCF-2; TCF2; VHNF1
UniProt ID: (Human) P35680
Entrez Gene ID: (Human) 6928
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.