Proteintech
Manufacturer Code:125331AP150UL
Catalog # 50-172-8046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HNF1B Fusion Protein Ag3232 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FJHN HNF 1 beta HNF 1B HNF1 homeobox B HNF1B HNF1beta HNF2 Homeoprotein LFB3 HPC11 LF B3 LFB3 MODY5 TCF 2 TCF2 Transcription factor 2 VHNF1; Hepatocyte nuclear factor 1-beta; hepatocyte nuclear factor-1 beta; HNF-1-beta; HNF-1B; HNF1 beta A; Homeoprotein LFB3; TCF-2; Transcription factor 2; Transcription factor 2 hepatic; Transcription factor 2 hepatic; LF-B3; variant hepatic nuclear factor; Transcription factor 2, hepatic; Transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor; Variant hepatic nuclear factor 1; vHNF1
Gene Aliases: FJHN; HNF-1-beta; HNF-1B; HNF-1Beta; HNF1B; HNF1beta; HNF2; HPC11; LF-B3; LFB3; MODY5; TCF-2; TCF2; VHNF1
UniProt ID: (Human) P35680, (Rat) P23899, (Mouse) P27889
Entrez Gene ID: (Human) 6928, (Rat) 25640, (Mouse) 21410
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.