R&D Systems
Manufacturer Code:PPH693900
Catalog # PPH693900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | H6939 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ39654 hepatic nuclear factor 4 alpha hepatocyte nuclear factor 4 alpha hepatocyte nuclear factor 4-alpha HNF4 alpha HNF4A HNF4a7 HNF-4-alpha HNF4alpha10/11/12 HNF4HNF4a8 MODY MODY1 NR2A1 NR2A1HNF4a9 NR2A21 Nuclear receptor subfamily 2 group A member 1 TCF TCF14 TCF-14 TCF14HNF4alpha Transcription factor 14 Transcription factor HNF-4 transcription factor-14; hepatic nuclear factor 4 alpha; hepatocyte nuclear factor 4, alpha; Hepatocyte nuclear factor 4-alpha; HNF-4-alpha; HNF4alpha10/11/12; Nuclear receptor subfamily 2 group A member 1; TCF-14; Transcription factor 14; Transcription factor HNF-4
Gene Aliases: FRTS4; HNF4; HNF4A; HNF4a7; HNF4a8; HNF4a9; HNF4alpha; MODY; MODY1; NR2A1; NR2A21; TCF; TCF14
UniProt ID: (Human) P41235
Entrez Gene ID: (Human) 3172
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.