R&D Systems
Manufacturer Code:FAB24001R100UG
Catalog # FAB2401R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2141A |
Immunogen | Human/Mouse/Rat HNF-3 beta/FoxA2 synthetic peptide Accession # Q9Y261 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rabbit Monoclonal Antibody
Protein Aliases: forkhead box A2 Forkhead box protein A2 FoxA2 hepatic nuclear factor-3-beta hepatocyte nuclear factor 3 beta hepatocyte nuclear factor 3-beta HNF3 beta HNF-3B HNF-3-beta HNF3BTCF-3B MGC19807 TCF3B Transcription factor 3B; Forkhead box protein A2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3, beta; Hepatocyte nuclear factor 3-beta; HNF-3-beta; HNF-3B; TCF-3B; Transcription factor 3B
Gene Aliases: FOXA2; HNF3B; TCF3B
UniProt ID: (Human) Q9Y261
Entrez Gene ID: (Human) 3170
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.