Novus Biologicals
Manufacturer Code:NB1001263
Catalog # NB1001263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-GVYSRPIMNSS corresponding to C-Terminus according to NP_710141.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: forkhead box A2 Forkhead box protein A2 hepatic nuclear factor-3-beta hepatocyte nuclear factor 3 beta hepatocyte nuclear factor 3-beta HNF-3B HNF-3-beta HNF3BTCF-3B MGC19807 TCF3B Transcription factor 3B; Forkhead box protein A2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3, beta; Hepatocyte nuclear factor 3-beta; HNF-3-beta; HNF-3B; TCF-3B; Transcription factor 3B
Gene Aliases: FOXA2; HNF3B; TCF3B
UniProt ID: (Human) Q9Y261
Entrez Gene ID: (Human) 3170
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.