R&D Systems
Manufacturer Code:MAB6778SP
Catalog # MAB6778SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 654126 |
Immunogen | <EM>E. coli</EM>-derived recombinant human HNF-3 alpha/FoxA1 Met253-Ser472 Accession # P55317 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: forkhead box A1 Forkhead box protein A1 FoxA1 hepatocyte nuclear factor 3 alpha hepatocyte nuclear factor 3-alpha HNF3 alpha HNF-3A HNF-3-alpha HNF3ATCF-3A MGC33105 TCF3A Transcription factor 3A; Forkhead box protein A1; Hepatocyte nuclear factor 3-alpha; HNF-3-alpha; HNF-3A; TCF-3A; Transcription factor 3A
Gene Aliases: FOXA1; HNF3A; TCF3A
UniProt ID: (Human) P55317
Entrez Gene ID: (Human) 3169
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.