Novus Biologicals
Manufacturer Code:NBP245269
Catalog # NBP245269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B11NB |
Immunogen | Partial recombinant human FOXA1 protein (between amino acids 350-500) [Uniprot: P55317]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: forkhead box A1 Forkhead box protein A1 hepatocyte nuclear factor 3 alpha hepatocyte nuclear factor 3-alpha HNF-3A HNF-3-alpha HNF3ATCF-3A MGC33105 TCF3A Transcription factor 3A; Forkhead box protein A1; Hepatocyte nuclear factor 3-alpha; HNF-3-alpha; HNF-3A; TCF-3A; Transcription factor 3A
Gene Aliases: FOXA1; HNF3A; TCF3A
UniProt ID: (Human) P55317
Entrez Gene ID: (Human) 3169
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.