Proteintech
Manufacturer Code:149141AP150UL
Catalog # 50-172-9830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HN1 Fusion Protein Ag6712 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Androgen regulated protein 2 ARM2 HN1 HN1A; Androgen-regulated protein 2; Hematological and neurological expressed 1 protein; hematological and neurological expressed sequence 1; Jupiter microtubule associated homolog 1; Jupiter microtubule associated homolog 1, N-terminally processed
Gene Aliases: ARM2; HN1; HN1A; JPT1
UniProt ID: (Human) Q9UK76, (Mouse) P97825, (Rat) Q6AXU6
Entrez Gene ID: (Human) 51155, (Mouse) 15374, (Rat) 287828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.