Invitrogen
Manufacturer Code:PA572623
Catalog # PIPA572623
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 132-160 amino acids from the Central region of DANRE hmx3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H6 family homeobox 3; homeo box (H6 family) 3; homeobox protein H6 family member 3; homeobox protein Nkx-5.1; Homeobox protein Nkx-5.1 Homeobox protein H6 family member 3 Homeobox protein HMX3 Hmx3 Nkx-5.1 Nkx5-1; transcription factor
Gene Aliases: hmx3; Nkx5-1; nkx5.1; zgc:109845
Entrez Gene ID: (Zebrafish) 60310
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.