Invitrogen
Manufacturer Code:PA569500
Catalog # PIPA569500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse HMX3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H6 family homeobox 3; H6 homeo box 3; homeo box (H6 family) 3; homeobox (H6 family) 3; Homeobox protein H6 family member 3; Homeobox protein HMX3; Homeobox protein Nkx-5.1; Homeobox protein Nkx-5.1 Homeobox protein H6 family member 3 Homeobox protein HMX3 Hmx3 Nkx-5.1 Nkx5-1; transcription factor
Gene Aliases: HMX3; NKX-5.1; NKX5-1; NKX5.1; RGD1559927; zgc:109845
UniProt ID: (Human) A6NHT5, (Mouse) P42581
Entrez Gene ID: (Horse) 100057903, (Human) 340784, (Dog) 611245, (Guinea pig) 100712708, (Mouse) 15373, (Rat) 293537, (Zebrafish) 60310
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.