Invitrogen
Manufacturer Code:PA538567
Catalog # PIPA538567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human HMG17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: high mobility group nucleosomal binding domain 2; High mobility group nucleosome-binding domain-containing protein 2; high mobility group protein 17; high mobility group protein N2; high-mobility group (nonhistone chromosomal) protein 17; high-mobility group nucleosomal binding domain 2; high-mobility group nucleosome-binding domain-containing protein 2; HMG17; Non-histone chromosomal protein HMG-17; nonhistone chromosomal protein HMG-17
Gene Aliases: HMG-17; HMG17; HMGN2
UniProt ID: (Human) P05204, (Mouse) P09602
Entrez Gene ID: (Human) 3151, (Mouse) 15331, (Rat) 114637
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.