Invitrogen
Manufacturer Code:PA575587
Catalog # PIPA575587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human HMGN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: High mobility group nucleosome-binding domain-containing protein 1; high mobility group protein 14; high-mobility group (nonhistone chromosomal) protein 14; high-mobility group nucleosome binding 1; high-mobility group nucleosome binding domain 1; high-mobility group nucleosome-binding domain-containing protein 1; Non-histone chromosomal protein HMG-14; Non-histone chromosomal protein HMG-14 High mobility group nucleosome-binding domain-containing protein 1 HMGN1 HMG14; nonhistone chromosomal protein HMG-14
Gene Aliases: HMG-14; HMG14; HMGN1
UniProt ID: (Human) P05114, (Mouse) P18608
Entrez Gene ID: (Human) 3150, (Mouse) 15312, (Rat) 360704
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.