Invitrogen
Manufacturer Code:PA568570
Catalog # PIPA568570
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human HMGN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ27265 FLJ31471 HMG14 MGC104230 MGC117425; High mobility group nucleosome-binding domain-containing protein 1; high mobility group protein 14; high-mobility group (nonhistone chromosomal) protein 14; high-mobility group nucleosome binding 1; high-mobility group nucleosome binding domain 1; high-mobility group nucleosome-binding domain-containing protein 1; Non-histone chromosomal protein HMG-14; nonhistone chromosomal protein HMG-14
Gene Aliases: HMG-14; HMG14; HMGN1
UniProt ID: (Bovine) P02316, (Human) P05114, (Mouse) P18608
Entrez Gene ID: (Horse) 100629274, (Pig) 100625838, (Bovine) 614915, (Human) 3150, (Rabbit) 108175971, (Dog) 608119, (Mouse) 15312, (Rat) 360704, (Guinea pig) 100725547
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.