Invitrogen
Manufacturer Code:PA585502
Catalog # PIPA585502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human HMGCS2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial); HMG-CoA synthase; HMGCS2 antibody HMG-CoA synthase antibody 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial) antibody hydroxymethylglutaryl-CoA synthase mitochondrial antibody 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial) antibody; Hydroxymethylglutaryl-CoA synthase, mitochondrial; testicular tissue protein Li 88
Gene Aliases: HMGCS2
UniProt ID: (Human) P54868
Entrez Gene ID: (Human) 3158
Molecular Function: lyase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.