Invitrogen
Manufacturer Code:PA5112573
Catalog # PIPA5112573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Hydroxymethylglutaryl-CoA synthase mitochondrial protein (426-508AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1300002P16 3-hydroxy-3-methylglutary-Coenzyme A synthase 2 3-hydroxy-3-methylglutaryl coenzyme A synthase 3-hydroxy-3-methylglutaryl-CoA synthase 2 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial) 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial) BOS_3051 hmgCoA HMG-CoA synthase Hmgcs1 Hmgcs2 hydroxymethylglutaryl-CoA synthase 2 Hydroxymethylglutaryl-CoA synthase mitochondrial mHS Mt3h3mg testicular tissue protein Li 88; 3-hydroxy-3-methylglutary-Coenzyme A synthase 2; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial); HMG-CoA synthase; hydroxymethylglutaryl-CoA synthase 2; Hydroxymethylglutaryl-CoA synthase, mitochondrial; testicular tissue protein Li 88
Gene Aliases: Hmgcs1; HMGCS2; Mt3h3mg
UniProt ID: (Human) P54868, (Rat) P22791
Entrez Gene ID: (Human) 3158, (Rat) 24450
Molecular Function:
lyase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.