Invitrogen
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | 1:10-1:50 |
Immunohistochemistry (Paraffin) (IHC (P)) | 1:10-1:50 |
Western Blot (WB) | 1:1,000 |
Host / Isotype | Rabbit / IgG |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 478-508 amino acids from the C-terminal region of human HMGCS2. |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | Lot-Specific |
Storage conditions | -20° C, Avoid Freeze/Thaw Cycles |
RRID | AB_2635043 |
The protein encoded by this gene belongs to the HMG-CoA synthase family. It is a mitochondrial enzyme that catalyzes the first reaction of ketogenesis, a metabolic pathway that provides lipid-derived energy for various organs during times of carbohydrate deprivation, such as fasting. Mutations in this gene are associated with HMG-CoA synthase deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.