Novus Biologicals
Manufacturer Code:NBP154995
Catalog # NBP154995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to HMGCS2(3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial)) The peptide sequence was selected from the C terminal of HMGCS2. Peptide sequence DLPKRLASRKCVSPEEFTEIMNQREQFYHKVNFSPPGDTNSLFPGTWY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl coenzyme A synthase 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial) 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial) EC 2.3.3.10 HMG-CoA synthase hydroxymethylglutaryl-CoA synthase mitochondrial mitochondrial 3-hydroxy-3-methylglutaryl-CoA synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial); HMG-CoA synthase; Hydroxymethylglutaryl-CoA synthase, mitochondrial; testicular tissue protein Li 88
Gene Aliases: HMGCS2
UniProt ID: (Human) P54868
Entrez Gene ID: (Human) 3158
Molecular Function:
lyase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.