Novus Biologicals
Manufacturer Code:NBP216861
Catalog # NBP216861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human HMGCS1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase; 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase 3-hydroxy-3-methylglutaryl coenzyme A synthase 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble) 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble) EC 2.3.3.10 HMG-CoA synthase HMGCS hydroxymethylglutaryl-CoA synthase cytoplasmic MGC90332; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble); HMG-CoA synthase; Hydroxymethylglutaryl-CoA synthase, cytoplasmic
Gene Aliases: HMGCS; HMGCS1
UniProt ID: (Human) Q01581
Entrez Gene ID: (Human) 3157
Molecular Function:
lyase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.