Invitrogen
Manufacturer Code:PA537367
Catalog # PIPA537367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length human HMGCR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 hydroxy 3 methylglutaryl Coenzyme A reductase 3 hydroxymethylglutaryl CoA reductase 3-hydroxy-3-methylglutaryl-coenzyme A reductase 3H3M HMDH HMG CoA reductase HMG CoAR HMG-CoA reductase Hmgcr Hydroxymethylglutaryl CoA reductase LDLCQ3 MGC103269; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl-CoA reductase; 3-hydroxy-3-methylglutaryl-coenzyme A reductase; HMG-CoA reductase; hydroxymethylglutaryl-CoA reductase
Gene Aliases: 3H3M; HMG-CoAR; HMGCR; LDLCQ3; Red
UniProt ID: (Human) P04035, (Mouse) Q01237, (Rat) P51639
Entrez Gene ID: (Human) 3156, (Mouse) 15357, (Rat) 25675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.