Invitrogen
Manufacturer Code:PA5115369
Catalog # PIPA5115369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HMGCL corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxy-3-methylglutarate-CoA lyase; 3-hydroxy-3-methylglutarate-CoA lyase 3-hydroxy-3-methylglutaryl-CoA lyase 3-hydroxy-3-methylglutaryl-Coenzyme A lyase 3-hydroxymethyl-3-methylglutaryl-CoA lyase 3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase AW476067 HL HMGCL HMG-CoA lyase hydroxymethylglutaricaciduria Hydroxymethylglutaryl-CoA lyase mitochondrial mitochondrial 3-hydroxy-3-methylglutaryl-CoA lyase; 3-hydroxy-3-methylglutaryl-CoA lyase; 3-hydroxy-3-methylglutaryl-Coenzyme A lyase; 3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase; HL; HMG-CoA lyase; hydroxymethylglutaricaciduria; Hydroxymethylglutaryl-CoA lyase, mitochondrial; mitochondrial 3-hydroxy-3-methylglutaryl-CoA lyase
Gene Aliases: AW476067; HL; HMGCL
UniProt ID: (Human) P35914, (Mouse) P38060, (Rat) P97519
Entrez Gene ID: (Human) 3155, (Mouse) 15356, (Rat) 79238
Molecular Function:
lyase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.