Invitrogen
Manufacturer Code:PA576431
Catalog # PIPA576431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human HMGB2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: High mobility group (nonhistone chromosomal) protein 2 high mobility group box 2 High mobility group protein 2 High mobility group protein B2 high-mobility group (nonhistone chromosomal) protein 2 HMG 2 HMG B2 HMG-2 HMG2 HMGB2; High mobility group protein 2; High mobility group protein B2; high-mobility group (nonhistone chromosomal) protein 2; high-mobility group box 2; HMG-2
Gene Aliases: C80539; HMG-2; HMG2; HMGB2
UniProt ID: (Human) P26583, (Rat) P52925, (Mouse) P30681
Entrez Gene ID: (Human) 3148, (Rat) 29395, (Mouse) 97165
Molecular Function:
DNA binding protein
HMG box transcription factor
chromatin/chromatin-binding protein
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.