Invitrogen
Manufacturer Code:PA514015
Catalog # PIPA514015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 92-118 amino acids from the central region of human HMGB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: High mobility group protein 2; High mobility group protein B2; High mobility group protein B2 High mobility group protein 2 HMG-2 synonyms: HMG2 name: HMGB2; high-mobility group (nonhistone chromosomal) protein 2; high-mobility group box 2; HMG-2
Gene Aliases: HMG2; HMGB2
UniProt ID: (Human) P26583
Entrez Gene ID: (Human) 3148
Molecular Function:
DNA binding protein
HMG box transcription factor
chromatin/chromatin-binding protein
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.