R&D Systems
Manufacturer Code:MAB1690SP
Catalog # MAB1690SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 115603 |
Immunogen | <EM>E. coli</EM>-derived recombinant human HMGB1/HMG-1 Gly2-Glu215 (predicted) Accession # P09429 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Amphoterin; Amphoterin high mobility group box 1 High mobility group protein 1 high mobility group protein B1 high-mobility group (nonhistone chromosomal) protein 1 high-mobility group box 1 HMG1 HMG-1 HMG1DKFZp686A04236 HMG3 SBP-1 Sulfoglucuronyl carbohydrate binding protein; High mobility group protein 1; High mobility group protein B1; high-mobility group (nonhistone chromosomal) protein 1; high-mobility group box 1; HMG-1; Sulfoglucuronyl carbohydrate binding protein
Gene Aliases: HMG-1; HMG1; HMG3; HMGB1; SBP-1
UniProt ID: (Human) P09429
Entrez Gene ID: (Human) 3146
Molecular Function:
DNA binding protein
HMG box transcription factor
chromatin/chromatin-binding protein
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.