Novus Biologicals
Manufacturer Code:NBP227117
Catalog # NBP227117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 100-166 of human HMGB1 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amphoterin; Amphoterin high mobility group box 1 High mobility group protein 1 high mobility group protein B1 high-mobility group (nonhistone chromosomal) protein 1 high-mobility group box 1 HMG-1 HMG1DKFZp686A04236 HMG3 SBP-1 Sulfoglucuronyl carbohydrate binding protein; High mobility group protein 1; High mobility group protein B1; high-mobility group (nonhistone chromosomal) protein 1; high-mobility group box 1; HMG-1; Sulfoglucuronyl carbohydrate binding protein
Gene Aliases: HMG-1; HMG1; HMG3; HMGB1; SBP-1
UniProt ID: (Human) P09429
Entrez Gene ID: (Human) 3146
Molecular Function: DNA binding protein HMG box transcription factor chromatin/chromatin-binding protein nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.