Invitrogen
Manufacturer Code:PA521320
Catalog # PIPA521320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 109 of HMGA2 (Uniprot IDNo.P52926) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BABL HMGI-C HMGIC LIPO STQTL9 High-mobility group protein HMGI-C high mobility group AT-hook protein 2 high mobility group protein HMGI-C high-mobility group (nonhistone chromosomal) protein isoform I-C; High mobility group AT-hook protein 2; High mobility group protein HMGI-C; high mobility group protein I, isoform C; high-mobility group (nonhistone chromosomal) protein isoform I-C; High-mobility group protein HMGI-C; non-histone chromosomal architectural protein HMGI-C
Gene Aliases: 9430083A20Rik; BABL; HMGA2; HMGI-C; HMGIC; LIPO; pg; pygmy; STQTL9
UniProt ID: (Human) P52926, (Mouse) P52927
Entrez Gene ID: (Human) 8091, (Mouse) 15364, (Rat) 84017
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.