R&D Systems
Manufacturer Code:AF3184SP
Catalog # AF3184SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human HMGA2 Ser2-Asp109 Accession # P52926 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BABL BABLHMGI-C high mobility group AT-hook 2 High mobility group AT-hook protein 2 high mobility group protein HMGI-C high-mobility group (nonhistone chromosomal) protein isoform I-C High-mobility group protein HMGI-C HMGIC HMGICSTQTL9 LIPO; High mobility group AT-hook protein 2; High mobility group protein HMGI-C; high-mobility group (nonhistone chromosomal) protein isoform I-C; High-mobility group protein HMGI-C
Gene Aliases: BABL; HMGA2; HMGI-C; HMGIC; LIPO; STQTL9
UniProt ID: (Human) P52926
Entrez Gene ID: (Human) 8091
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.