Novus Biologicals
Manufacturer Code:NBP136744
Catalog # NBP136744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SQPLASKQEKDGT corresponding to internal region (near N-Terminus) according to NP_002122.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: high mobility group AT-hook 1 High mobility group AT-hook protein 1 High mobility group protein A1 high mobility group protein HMG-I/HMG-Y High mobility group protein R high-mobility group (nonhistone chromosomal) protein isoforms I and Y HMGA1A HMG-I(Y) HMGIYMGC12816 HMG-R MGC4242 MGC4854 nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y; High mobility group AT-hook protein 1; High mobility group protein A1; High mobility group protein HMG-I/HMG-Y; High mobility group protein R; high-mobility group (nonhistone chromosomal) protein isoforms I and Y; HMG-I(Y); nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y
Gene Aliases: HMG-R; HMGA1; HMGA1A; HMGIY
UniProt ID: (Human) P17096
Entrez Gene ID: (Human) 3159
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.