Invitrogen
Manufacturer Code:PA141326
Catalog # 01-675-663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 19-32 of human HMGa1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: High mobility group AT-hook protein 1; high mobility group HMGA1A; high mobility group HMGA1B; high mobility group protein; high mobility group protein 1A; High mobility group protein A1; High mobility group protein HMG-I/HMG-Y; high mobility group protein I; High mobility group protein R; high-mobility group (nonhistone chromosomal) protein isoforms I and Y; HMG-I(Y); HMGA1; non-histone protein; non-histone protein; HMGA1A; non-histone protein; HMGA1B; nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y
Gene Aliases: AL023995; HMG-I(Y); HMG-R; HMGA1; HMGA1A; Hmga1b; Hmgi; HMGIY; Hmgy
UniProt ID: (Human) P17096, (Dog) Q6URC2, (Mouse) P17095, (Rat) Q8K585
Entrez Gene ID: (Human) 3159, (Pig) 100157848, (Dog) 442946, (Mouse) 15361, (Rat) 117062
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.