Novus Biologicals
Manufacturer Code:NBP245986
Catalog # NBP245986
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H4 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 1-281 of human HMG20A(NP_060670) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ10739 high-mobility group 20A HMG box domain containing 1 HMG box-containing protein 20A HMG domain-containing protein 1 HMGX1HMG domain-containing protein HMGX1 HMGXB1high mobility group protein 20A; High mobility group protein 20A; HMG box domain containing 1; HMG box-containing protein 20A; HMG domain-containing protein 1; HMG domain-containing protein HMGX1
Gene Aliases: HMG20A; HMGX1; HMGXB1
UniProt ID: (Human) Q9NP66
Entrez Gene ID: (Human) 10363
Molecular Function:
DNA binding protein
HMG box transcription factor
chromatin/chromatin-binding protein
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.