Novus Biologicals
Manufacturer Code:NBP136961
Catalog # NBP136961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-ADPSPQNSTADTSK corresponding to internal region according to NP_000850.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl CoA reductase (NADPH) 3-hydroxy-3-methylglutaryl-CoA reductase 3-hydroxy-3-methylglutaryl-Coenzyme A reductase EC 1.1.1 EC 1.1.1.34 HMG-CoA reductase hydroxymethylglutaryl-CoA reductase LDLCQ3; 3-hydroxy-3-methylglutaryl-coenzyme A reductase; HMG-CoA reductase; hydroxymethylglutaryl-CoA reductase
Gene Aliases: HMGCR; LDLCQ3
UniProt ID: (Human) P04035
Entrez Gene ID: (Human) 3156
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.