OriGene
Manufacturer Code:CF802669
Catalog # 50-166-6093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F4 |
Immunogen | Full length human recombit protein of human HMBS produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HMBS; Hydroxymethylbilane synthase; PBG-D; PBG-D PBGD PORC UPS; Porphobilinogen deaminase; porphyria, acute; Chester type; Pre-uroporphyrinogen synthase; uroporphyrinogen I synthase; uroporphyrinogen I synthetase
Gene Aliases: HMBS; PBG-D; PBGD; PORC; UPS
UniProt ID: (Human) P08397
Entrez Gene ID: (Human) 3145
Molecular Function: deaminase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.