Invitrogen
Manufacturer Code:PA552253
Catalog # PIPA552253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HMBS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative name: porphobilinogen deaminase; HEM3 HMBS Hydroxymethylbilane synthase PBG D PBG-D PBGD Porphobilinogen deaminase Pre uroporphyrinogen synthase Pre-uroporphyrinogen synthase UPS Uroporphyrinogen I synthase Uroporphyrinogen I synthetase; HMBS; Hydroxymethylbilane synthase; PBG-D; Porphobilinogen deaminase; porphyria, acute; Chester type; Pre-uroporphyrinogen synthase; uroporphyrinogen I synthase; uroporphyrinogen I synthetase
Gene Aliases: hemC; HMBS; PBG-D; PBGD; PORC; T25658; UPS; URO-S; Uros1
UniProt ID: (Human) P08397, (Mouse) P22907
Entrez Gene ID: (Human) 3145, (Mouse) 15288, (Rat) 25709
Molecular Function:
deaminase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.