Invitrogen
Manufacturer Code:PA544169
Catalog # PIPA544169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human HMBS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative name: porphobilinogen deaminase; HMBS; Hydroxymethylbilane synthase; Hydroxymethylbilane synthase PBG-D PBGD UPS PORC; PBG-D; Porphobilinogen deaminase; porphyria, acute; Chester type; Pre-uroporphyrinogen synthase; uroporphyrinogen I synthase; uroporphyrinogen I synthetase
Gene Aliases: hemC; HMBS; PBG-D; PBGD; PORC; T25658; UPS; URO-S; Uros1
UniProt ID: (Human) P08397, (Bovine) Q2KIN5, (Mouse) P22907
Entrez Gene ID: (Rabbit) 100341318, (Human) 3145, (Dog) 489373, (Sheep) 100528012, (Bovine) 515614, (Horse) 100071339, (Guinea pig) 100720802, (Mouse) 15288, (Rat) 25709
Molecular Function: deaminase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.